missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD6B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93357-0.1ml
This item is not returnable.
View return policy
Description
PARD6B Polyclonal antibody specifically detects PARD6B in Human samples. It is validated for Western Blot
Specifications
| PARD6B | |
| Polyclonal | |
| Western Blot 1:1000-1:4000 | |
| par-6 (partitioning defective 6, C.elegans) homolog beta, PAR-6 beta, par-6 partitioning defective 6 homolog beta (C. elegans), PAR6B, PAR-6B, partitioning defective 6 homolog beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 253-372 of human PARD6B (NP_115910.1). NNVVRNSRTSGSSGQSTDNSLLGYPQQIEPSFEPEDEDSEEDDIIIEDNGVPQQIPKAVPNTESLESLTQIELSFESGQNGFIPSNEVSLAAIASSSNTEFETHAPDQKLLEEDGTIITL | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 84612 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction