missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PARD6G Polyclonal Antibody
GREENER_CHOICE

Product Code. 13271699
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13271699 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13271699 Supplier Invitrogen™ Supplier No. PA524684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PARD6G Polyclonal Antibody for Western Blot, IHC (P)

PAR6G involved in asymmetrical cell division and cell polarization processes. PAR6G may play a role in the formation of epithelial tight junctions. The PARD6-PARD3 complex links GTP-bound Rho small GTPases to atypical protein kinase C proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD6G
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PARD6G
Gene Accession No. Q9BYG4
Gene Alias 2410049N21Rik; par-6 family cell polarity regulator gamma; PAR-6 gamma; PAR-6 gamma protein; par-6 partitioning defective 6 homolog gamma; Par6a; PAR6D; PAR6G; PAR-6G; PAR6gamma; Pard6g; Partitioning defective 6 homolog gamma; partitioning-defective protein 6, alpha
Gene Symbols PARD6G
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 18-47 amino acids from the N-terminal region of human PAR6G.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.