missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parecoxib Sodium, MedChemExpress
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Description
Parecoxib Sodium (SC 69124A) is a highly selective and orally active COX-2 inhibitor, the prodrug of Valdecoxib (HY-15762). Parecoxib Sodium is a nonsteroidal anti-inflammatory agent (NSAID) and inhibits prostaglandin (PG) synthesis. Parecoxib Sodium can be used for the relief of acute postoperative pain and symptoms of chronic inflammatory conditions such as osteoarthritis and rheumatoid arthritis in vivo.
Specifications
Specifications
| Chemical Name or Material | Parecoxib Sodium |
| CAS | 198470-85-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H17N2NaO4S |
| Quantity | 100 mg |
| Synonym | SC 69124A |
| Solubility Information | DMSO : ≥ 100 mg/mL (254.84 mM) ∣H2O : ≥ 100 mg/mL (254.84 mM) |
| SMILES | CCC(N([Na])S(=O)(C1=CC=C(C2=C(C)ON=C2C3=CC=CC=C3)C=C1)=O)=O |
| Molecular Weight (g/mol) | 392.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction