missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parecoxib Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Parecoxib Sodium
Specifications
Specifications
| Chemical Name or Material | Parecoxib sodium |
| CAS | 198470-85-8 |
| Analyte or Component Names | Parecoxib sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H17 N2 O4 S . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Propanamide, N-[[4-(5-methyl-3-phenyl-4-isoxazolyl)phenyl]sulfonyl]-, sodium salt (1:1), Propanamide, N-[[4-(5-methyl-3-phenyl-4-isoxazolyl)phenyl]sulfonyl]-, sodium salt (9CI), Dynastat, Parecoxib sodium, Rayzon, SC 69124A |
| InChI Formula | InChI=1S/C19H18N2O4S.Na/c1-3-17(22)21-26(23,24)16-11-9-14(10-12-16)18-13(2)25-20-19(18)15-7-5-4-6-8-15;/h4-12H,3H2,1-2H3,(H,21,22);/q;+1/p-1 |
| SMILES | [Na+].CCC(=O)[N-]S(=O)(=O)c1ccc(cc1)c2c(C)onc2c3ccccc3 |
| IUPAC Name | sodium;[4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)phenyl]sulfonyl-propanoylazanide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction