missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Antibody (4H4) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-92715JF525
This item is not returnable.
View return policy
Description
Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Bovine,Human,Mouse (Negative),Rat (Negative) samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Gene Knock-Out,Immunocytochemistry/ Immunofluorescence
Specifications
| Park7/DJ-1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497] | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 11315 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Simple Western, Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| 4H4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction