missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-270AF488
This item is not returnable.
View return policy
Description
Park7/DJ-1 Polyclonal antibody specifically detects Park7/DJ-1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In vitro Assay
Specifications
| Park7/DJ-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 488 | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| A synthetic peptide corresponding to the C-terminus (within residues 150-189) of human Park7(DJ-1). [UniProt# Q99497] | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 11315 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction