Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Bulkhead Union
Click to view available options
Tubing Size:
1/4 in.
1/8 in.
Unit Size:
Each
Description
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
| Description | Compression Connection |
| Autoclavable | Autoclavable |
| Material | PFA |
| Tubing Size | 1/8 in. |
| Outlet Port | 1/8 x 1/8 in. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Color | Clear |
| Product Type | Compression Bulkhead Union |