missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parkin Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41287MFV450
This item is not returnable.
View return policy
Description
Parkin Polyclonal antibody specifically detects Parkin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Parkin | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human PARK2. The immunogen is located within amino acids 110 - 160 of PARK2. Amino Acid Squence: HTDSRKDSPPAGSPAG | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 5071 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction