missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Parkin Monoclonal Antibody (PRK109)
GREENER_CHOICE

Product Code. 10797803
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10797803 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10797803 Supplier Invitrogen™ Supplier No. 390900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Parkin Monoclonal Antibody for ELISA, ICC/IF, IP, Western Blot

The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parkin
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PRK109
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PRKN
Gene Accession No. O60260, Q9JK66, Q9WVS6
Gene Alias AAN12155.1; AR-JP; CG10523; CG10523-PB; CG10523-PC; Dmel\CG10523; Dmel_CG10523; Dpark; dParkin; D-parkin; dpk; E3 ubiquitin ligase; E3 ubiquitin-protein ligase parkin; LPRS2; OTTHUMP00000017562; OTTHUMP00000017563; OTTHUMP00000017564; park; park1; PARK2; Parkin; parkin 2; parkin F; Parkin isoform 1; parkin protein; parkin RBR E3 ubiquitin protein ligase; Parkin RBR E3 ubiquitin-protein ligase; parkin variant SV5DEL; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin); park-PB; park-PC; PDJ; PK; Prkn; SD01679
Gene Symbols PRKN
Host Species Mouse
Immunogen Full length recombinant human Parkin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5071, 50873, 56816
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.