Learn More
PARL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26679-1-AP-20UL
 View more versions of this product
Product Code. 16865075
  • € 145.00 / 20µL
Description
PARL is a mitochondrial integral membrane protein and a member of the highly conserved rhomboid superfamily of membrane proteins. PARL is required for the control of apoptosis during postnatal growth and is required for the processing of OPA1, a protein that prevents the release of cytochrome C from the mitochondria in response to apoptotic signals. In lymphocytes and neurons, PARL′s association with other proteins, such as the Bcl-2 family-related protein Hax1 and Omi can lead to the proteolytic processing of Omi by PARL, preventing the accumulation of Bax, thereby suppressing apoptosis. Variations in this gene have been associated with increased risk for type 2 diabetes. PARL may also play a role in the progression of Alzheimer′s disease through its associations with presenilin-1 and -2. Multiple isoforms of PARL are known to exist.
Specifications
Specifications
PARL
Polyclonal
Unconjugated
PARL
PARL, Pbeta, PRO2207, PSARL, PSARL1, PSENIP2, RHBDS1
Rabbit
Antigen Affinity Chromatography
RUO
55486
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H300
PARL
PARL Fusion Protein Ag24789