missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARN Antibody (OTI4D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46321
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
PARN Monoclonal specifically detects PARN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PARN | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| DANEC 3.1.13.4, Deadenylating nuclease, Deadenylation nuclease, poly(A)-specific ribonuclease (deadenylation nuclease), poly(A)-specific ribonuclease PARN, Polyadenylate-specific ribonuclease | |
| Mouse | |
| 73.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4D12 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| PARN | |
| Human recombinant protein fragment corresponding to amino acids 1-266 of human PARN (NP_002573) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 5073 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction