missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARN Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802653S
This item is not returnable.
View return policy
Description
PARN Monoclonal antibody specifically detects PARN in Human samples. It is validated for Western BlotSpecifications
| PARN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95453 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5073 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4C9 | |
| Unconjugated | |
| Parn | |
| 1200003I18Rik; DAN; Deadenylating nuclease; deadenylation nuclease; DKCB6; Parn; PFBMFT4; poly(A)-specific ribonuclease; poly(A)-specific ribonuclease (deadenylation nuclease); poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease; RGD1565449 | |
| Human recombinant protein fragment corresponding to amino acids 1-266 of human PARN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction