missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111353
This item is not returnable.
View return policy
Description
Immunogen sequence: LVGPMEITMG SLEKAGPVSP GCVKLAGQEG LVEMVLLMEP GAMRFLQLYH EDLLAGLGDV ALLPLEGPDM TGFRLCG Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 62%.
Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors. Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors (Yu et al., 2005).
Specifications
| PARP10 | |
| Polyclonal | |
| Unconjugated | |
| PARP10 | |
| ADP-ribosyltransferase diphtheria toxin-like 10; ARTD10; FLJ14464; LOC102548778; Parp10; PARP-10; poly (ADP-ribose) polymerase family, member 10; poly [ADP-ribose] polymerase 10; poly [ADP-ribose] polymerase 10-like; poly(ADP-ribose) polymerase family member 10; Protein mono-ADP-ribosyltransferase PARP10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84875 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.90 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53GL7 | |
| PARP10 | |
| Recombinant Protein corresonding to Human PARP10. Recombinant protein control fragment (Product #RP-104946). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction