missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP14 Recombinant Rabbit Monoclonal Antibody (HL1178)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546948
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Poly [ADP-ribose] polymerase 14 (PARP14) is a B aggressive lymphoma protein belonging to the PARP protein family. Poly ADP ribosylation is a DNA damage dependent post translational modification of histones and other nuclear proteins that mediates DNA repair. PARP proteins with a macrodomain are also involved in gene expression through transcriptional regulation. PARP14 controls STAT6 dependent transcription by enhancing binding to the promoter in the presence of a positive signal (i.e. IL4 stimulation).
Specifications
| PARP14 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q2EMV9, Q460N5 | |
| PARP14 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PARP14. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1178 | |
| Unconjugated | |
| PARP14 | |
| 1600029O10Rik; ADP-ribosyltransferase diphtheria toxin-like 8; ARTD8; B aggressive lymphoma protein 2; B-aggressive lymphoma 2; BAL2; BC021340; CoaSt6; Collaborator of STAT6; KIAA1268; mKIAA1268; Parp14; PARP-14; pART8; poly (ADP-ribose) polymerase family, member 14; poly [ADP-ribose] polymerase 14; poly(ADP-ribose) polymerase family member 14; Protein mono-ADP-ribosyltransferase PARP14; protein mono-ADP-ribosyltransferase PARP14; poly [ADP-ribose] polymerase 14; RGD1310490; Unknown (protein for IMAGE:8227979) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 303903, 54625, 547253 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction