missing translation for 'onlineSavingsMsg'
Learn More

PARP16 Mouse anti-Human, Clone: OTI2C8, liquid, TrueMAB™

Product Code. 17571461
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17571461 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17571461 Supplier Origene Technologies Supplier No. TA500452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The function of this protein has not been specifically defined.

Specifications

Antigen PARP16
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2C8
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PARP16
Gene Accession No. Q8N5Y8
Gene Alias ARTD15, C15orf30, pART15
Gene Symbols PARP16
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PARP16 expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54956
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.