missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP2 Antibody - Unpurified, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-185SS
This item is not returnable.
View return policy
Description
PARP2 Polyclonal specifically detects PARP2 in Human samples. It is validated for Western Blot.
Specifications
| PARP2 | |
| Polyclonal | |
| Western Blot 1:250-1:500 | |
| ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2, ADPRT-2, ADPRT2pADPRT-2, ADPRTL2EC 2.4.2.30, ADPRTL3, hPARP-2, NAD(+) ADP-ribosyltransferase 2, PARP-2, poly (ADP-ribose) polymerase 2, poly (ADP-ribose) polymerase family, member 2, poly (ADP-ribosyl) transferase-like 2, poly [ADP-ribose] polymerase 2, poly(ADP-ribose) synthetase, Poly[ADP-ribose] synthase 2, poly[ADP-ribose] synthetase 2 | |
| Rabbit | |
| 66 kDa | |
| 0.025 mL | |
| Primary | |
| N-terminus of human PARP 2 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| PARP2 | |
| Residues 43-59 [QRQESKKMPVAGGKANK] of the 62 kDa human PARP-2 protein. | |
| Whole Antiserum | |
| Apoptosis, DNA Repair, Hypoxia | |
| 10038 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction