missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PARP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17177408 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177408 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177408 Supplier Invitrogen™ Supplier No. PA5116175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PARP2 Polyclonal Antibody for Western Blot

This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PARP2
Gene Accession No. O88554
Gene Alias ADP-ribosyltransferase (NAD+, poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT2; ADPRT-2; ADPRTL2; ADPRTL3; ARTD2; Aspartl2; C78626; DNA ADP-ribosyltransferase PARP2; HGNC:272; hPARP-2; mPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; parp 2; PARP2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; poly [ADP-ribose] polymerase 2; poly(ADP-ribose) polymerase 2; poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2
Gene Symbols PARP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 306-336 amino acids from the C-terminal region of human PARP2.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11546
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.