missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540773
This item is not returnable.
View return policy
Description
Peptide sequence: LLDLFEVEKD GEKEAFREDL HNRMLLWHGS RMSNWVGILS HGLRIAHPEA Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 92%.
This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.
Specifications
| PARP2 | |
| Polyclonal | |
| Unconjugated | |
| PARP2 | |
| ADP-ribosyltransferase (NAD+, poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT2; ADPRT-2; ADPRTL2; ADPRTL3; ARTD2; Aspartl2; C78626; DNA ADP-ribosyltransferase PARP2; HGNC:272; hPARP-2; mPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; parp 2; PARP2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; poly [ADP-ribose] polymerase 2; poly(ADP-ribose) polymerase 2; poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10038, 290027 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UGN5 | |
| PARP2 | |
| Synthetic peptide directed towards the middle region of human PARP2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction