missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARS2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94594-0.1ml
This item is not returnable.
View return policy
Description
PARS2 Polyclonal antibody specifically detects PARS2 in Human samples. It is validated for Western Blot
Specifications
| PARS2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp727A071, EC 6.1.1.15, MGC14416, MGC19467, MT-PRORS, probable prolyl-tRNA synthetase, mitochondrial, proline tRNA ligase 2, mitochondrial (putative), Proline--tRNA ligase, prolyl-tRNA synthetase (mitochondrial)(putative), prolyl-tRNA synthetase 2, mitochondrial (putative), prolyl-tRNA synthetase, mitochondrial (putative), proRS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-220 of human PARS2 (NP_689481.2). HHCAPRRGRRLLLSRVFQPQNLREDRVLSLQDKSDDLTCKSQRLMLQVGLIYPASPGCYHLLPYTVRAMEKLVRVIDQEMQAIGGQKVNMPSLSPAELWQATNRWDLMGKELLRLRDRHGKEYCLGPTHEEAITALIASQKKLSYKQLPFLLYQVTRKFRDEPRPRFGLLRGREFYMKDMYTFDSSPEAAQ | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 25973 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction