Learn More
Amarell™ Partial Immersion Toluene Filled Calibrated Thermometers Acc. to ASTM
Precision thermometers for special applications, each manufactured as per the lastest specifications of the ASTM-Standard E1.
Brand: Amarell™ A300090
Additional Details : Weight : 0.02800kg
Description
- Round solid stem
- Toluene filling
- Durable pigment
- Finish top round
- In transparent square plastic case
- Suitable for government verification or calibration
Specifications
6C | |
Low cloud and pour | |
Toluene, White Capillary Back | |
Round stem | |
76 mm |
ASTM | |
Partial Immersion | |
ASTM Thermometer | |
Yes |