Learn More
Amarell™ Partial Immersion Toluene Filled Calibrated Thermometers Acc. to ASTM
Precision thermometers for special applications, each manufactured as per the lastest specifications of the ASTM-Standard E1.
Brand:  Amarell™ A300090
Additional Details : Weight : 0.02800kg
Product Code. 10298010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Round solid stem
  • Toluene filling
  • Durable pigment
  • Finish top round
  • In transparent square plastic case
  • Suitable for government verification or calibration
Specifications
Specifications
6C
Low cloud and pour
Toluene, White Capillary Back
Round stem
76 mm
ASTM
Partial Immersion
ASTM Thermometer