Learn More
PARVA Monoclonal Antibody (OTI3H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506003S
Encompass_Preferred
 View more versions of this product
Product Code. 17555232
  • € 242.00 / 30µL
Description
PARVA Monoclonal antibody specifically detects PARVA in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
PARVA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9EPC1, Q9HB97, Q9NVD7
Mouse
Affinity chromatography
RUO
55742, 57341, 57342
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI3H9
Unconjugated
PARVA
2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha
Full length human recombinant protein of human PARVA produced in HEK293T cell.