missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARVA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531182
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, mouse kidney. Predicted reactivity: Mouse (99%), Rat (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| PARVA | |
| Polyclonal | |
| Unconjugated | |
| PARVA | |
| 2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55742, 57342 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9EPC1, Q9NVD7 | |
| PARVA | |
| Recombinant protein encompassing a sequence within the center region of human Parvin alpha. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction