Learn More
PARVA Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55268-1-AP-20UL
 View more versions of this product
Product Code. 16836765
  • € 145.00 / 20µL
Description
This antibody is specific to PARVA.
The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
Specifications
PARVA
Polyclonal
Unconjugated
PARVA
Actopaxin, Alpha parvin, CH ILKBP, MXRA2, PARVA, parvin, alpha
Rabbit
Antigen Affinity Chromatography
RUO
476859, 55742, 57341, 57342
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPC1, Q9HB97, Q9NVD7
PARVA
Peptide
20 μL
Primary
Canine, Human, Mouse, Rat