missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody (3C9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50038AF405
This item is not returnable.
View return policy
Description
Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Parvalbumin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 3C9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Full-length recombinant human Parvalbumin expressed in and purified from E. coli. [UniProt# P20472] | |
| 0.1 mL | |
| Neuroscience | |
| 5816 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction