Learn More
Parvalbumin Antibody (3C9), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50038V
Product Code. 18744030
  • ONLINE EXCLUSIVE PRICE
    € 614.00
    € 580.65 / 0.10mL
    Save € 33.35
    5% Off
Description
Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Parvalbumin
Monoclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
3C9
0.1 mL
Store at 4°C in the dark.