missing translation for 'onlineSavingsMsg'
Learn More

Parvalbumin Antibody (3C9), Novus Biologicals™

Product Code. 18667898 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667898 0.025 mL 0.03mL
18647898 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18667898 Supplier Novus Biologicals Supplier No. NBP250038SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parvalbumin
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3C9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000
Formulation 50% PBS, 50% glycerol
Gene Accession No. P20472, P20472
Gene Alias D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV
Host Species Mouse
Immunogen Full-length recombinant human Parvalbumin expressed in and purified from E. coli. [UniProt# P20472]
Purification Method Immunogen affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5816
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.