missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody, DyLight 650 , Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97031C
This item is not returnable.
View return policy
Description
Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Parvalbumin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Produced in rabbits immunized with purified, recombinant Human Parvalbumin (P20472; Met1-Ser110). | |
| 0.1 mL | |
| Neuroscience | |
| 5816 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction