missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97030PE
This item is not returnable.
View return policy
Description
Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human samples. It is validated for ELISA
Specifications
| Parvalbumin | |
| Polyclonal | |
| ELISA | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Produced in rabbits immunized with purified, recombinant Human Parvalbumin (Accession#: P20472; Met1-Ser110) | |
| 0.1 mL | |
| Neuroscience | |
| 5816 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction