missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Parvalbumin Polyclonal Antibody

Chicken Polyclonal Antibody

Brand:  Invitrogen™ PA5143579

 View more versions of this product

Product Code. 17856627

  • € 504.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Parvalbumin Polyclonal Antibody for ICC/IF, IHC (P), IHC (PFA), Western Blot

Parvalbumin (PV) is a calcium binding protein expressed in specific muscle fibers and fast-firing neurons. PV consists of a single, unbranched chain of linked amino acids and belongs to a larger group of EF hand proteins. Studies have demonstrated that parvalbumin acts in the decay of calcium in the contraction/ relaxation cycle of fast twitch muscles. This data has shown a positive correlation between the rate of relaxation and the concentration of parvalbumin. Parvalbumin is also expressed in a specific population of GABAergic interneurones which are thought to play a role in maintaining the balance between excitation and inhibition in the cortex as well as the hippocampus. In amyotrophic lateral sclerosis (ALS) patents, parvalbumin immunoreactivity is specifically absent from neuron populations lost early in ALS.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Parvalbumin
Polyclonal
Unconjugated
Pvalb
D22S749; HGNC:9704; MGC116759; PALB1; Parv; parvalbumin; Parvalbumin (calcium binding protein); parvalbumin alpha; PV; Pva; PVALB
Chicken
100 μL
Primary
Human, Mouse, Rat
Antibody
IgY
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Conc. Not Determined
PBS with 0.02% sodium azide
P02625, P20472, P32848
Pvalb
Full-length recombinant human protein expressed in and purified from E. coli.
RUO
19293, 25269, 5816
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.