missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARVB Antibody (OTI1B7), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73248C
This item is not returnable.
View return policy
Description
PARVB Monoclonal specifically detects PARVB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PARVB | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PARVB | |
| Full length recombinant protein of human PARVB (NP_037459) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B7 | |
| Western Blot, Immunohistochemistry | |
| affixin, beta-parvin, CGI-56, parvin, beta | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 29780.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction