Learn More
TG Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804834S
Encompass_Preferred
 View more versions of this product
Product Code. 17564563
  • € 242.00 / 30µL
Description
TG Monoclonal antibody specifically detects TG in Human samples. It is validated for Western Blot
Specifications
Specifications
TG
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P50221
Mouse
Affinity chromatography
RUO
4222
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G4
Unconjugated
MEOX1
AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor
Full length human recombinant protein of human MEOX1 produced in E.coli.