missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARVB Monoclonal Antibody (OTI3G1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526197
This item is not returnable.
View return policy
Description
PARVB Monoclonal Antibody for Western Blot, IHC (P)
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
Specifications
| PARVB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HBI1 | |
| Parvb | |
| Full length human recombinant protein of PARVB produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G1 | |
| Unconjugated | |
| Parvb | |
| Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29780 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction