missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARVB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140253
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VVRKREGLLH SSHISEELTT TTEMMMGRFE Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts. Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.
Specifications
| PARVB | |
| Polyclonal | |
| Unconjugated | |
| Parvb | |
| Affixin; AI595373; AW742462; beta-parvin; CGI-56; D15Gsk1; fd02e01; hypothetical protein LOC325686; Parvb; parvin beta; parvin, beta; wu:fd02e01; zgc:73117 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBI1 | |
| Parvb | |
| Recombinant protein fragment of Human PARVB (Product #RP-107177) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction