missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PARVG Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110073

Product Code. 17158003

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MEPEFLYDLL QLPKGVEPPA EEELSKGGKK KYLPPTSRKD PKFEELQKVL MEWINATLLP EHIVVRSLEE DMFDGLILHH LFQRLAALKL EAEDIALTAT SQKHKLTVVL EAVNRSLQLE EWQAKWSVES IFNKDLLSTL HLLVALAKRF QPDLSLPTNV QVEVITIEST KSGLKSEKLV EQLTEYSTDK DEPPKDVFDE LFKLAPEKVN AVKEAIVNFV NQKLDRLGLS VQNLDTQFAD GVILLLLIGQ LEGFFLHLKE FYLTPNSPAE MLHNVTLALE LLKDEGLLSC PVSPEDIVNK DAKSTLRVLY GLFCKHTQKA HRDRTPHGAP N.

Probably plays a role in the regulation of cell adhesion and cytoskeleton organization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PARVG
Polyclonal
Unconjugated
Parvg
AI413459; Gamma-parvin; gamma-parvin; LOW QUALITY PROTEIN: gamma-parvin; PARVG; parvin gamma; parvin, gamma; zgc:162304
Rabbit
Affinity chromatography
RUO
64099
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.17 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q9ERD8
Parvg
Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human PARVG (NP_071424.1).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.