missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvin gamma Antibody (PARVG/6312R), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08729AF594
This item is not returnable.
View return policy
Description
Parvin gamma Monoclonal antibody specifically detects Parvin gamma in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Parvin gamma | |
| Monoclonal | |
| Alexa Fluor 594 | |
| gamma-parvin, parvin, gamma | |
| GST-tagged recombinant protein corresponding to human Parvin gamma (Uniprot: Q9HBI0) | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 64098 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PARVG/6312R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction