missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASD1 Antibody (2ALCC128), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50234AF405
This item is not returnable.
View return policy
Description
PASD1 Monoclonal specifically detects PASD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
| PASD1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PASD1 | |
| GST-PASD1 recombinant protein containing the 540-773 aas of PASD1, a region only present in PASD1b. | |
| 0.1 mL | |
| Epigenetics | |
| 139135 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| 2ALCC128 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific for the recombinant protein (aa 540-773) present only in the longer PASD1b protein (C-terminus) | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction