missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASD1 Antibody (3C1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00139135-M08
This item is not returnable.
View return policy
Description
PASD1 Monoclonal antibody specifically detects PASD1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| PASD1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1 | |
| PASD1 (NP_775764, 1 a.a. ∽ 100 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLL | |
| 0.1 mg | |
| Epigenetics | |
| 139135 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 3C1 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_775764 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction