missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASK Antibody (OTI8G9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02242
This item is not returnable.
View return policy
Description
PASK Monoclonal specifically detects PASK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PASK | |
| Monoclonal | |
| 0.93 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| PASK | |
| Full length human recombinant protein of human PASK (NP_055963) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI8G9 | |
| Unconjugated | |
| DKFZp434O051, EC 2.7.11.1, hPASK, KIAA0135DKFZp686P2031, PAS domain containing serine/threonine kinase, PAS-kinase, PASKINPAS domain-containing serine/threonine-protein kinase, PAS-serine/threonine kinase, STK37 | |
| Mouse | |
| 142.7 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 23178 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction