Learn More
Hecht Karl™ Pasteur Pipets
Product Code. 10727322
Click to view available options
Length (Metric):
150 mm
230 mm
Type:
Long Form
Short Form
Unit Size:
Pack of 250
Product Code. 10727322
Brand: Hecht Karl™ 40567001
Only null left
Add to basket
Edge
Add to basket
Manufactured from glass for liquids susceptible to leaching from plastic pipets. Karl Hecht™ Pasteur Pipets are ideal for transferring small quantities of liquid.
  • Uniform shape
  • Disposable
  • Constriction in upper section for cotton plug 
Specifications
Diameter (Metric) Inner Tip Opening: 1 mm
Diameter (Metric) Outer 7 mm
Quantity 250 Pack
Length (Metric) 150 mm
Type Short Form