missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched 1/PTCH Antibody (OTI5C7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73253AF350
This item is not returnable.
View return policy
Description
Patched 1/PTCH Monoclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Patched 1/PTCH | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PTCH1 | |
| Recombinant protein fragment corresponding to amino acids 122-436 of human Patched 1/PTCH (NP_000255) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5C7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4′, PTCH protein -10, PTCH11 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| 5727.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction