missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23623PCP
This item is not returnable.
View return policy
Description
Patched Polyclonal antibody specifically detects Patched in Mouse samples. It is validated for Western Blot
Specifications
| Patched | |
| Polyclonal | |
| Western Blot | |
| PTCH | |
| Reacts with residues 269-279 [KKINYQVDSWE] of the mouse PTCH protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction