Learn More
DiscoverX PathHunter™ Anti-EA Antibody
The PathHunter™ Anti-EA Antibody is an antibody against the large enzyme acceptor (EA) fragment of β-galactosidase. This antibody is provided as a vial of 100μg of antibody in buffer at 2mg/mL.
Brand: DiscoverX 93-0246L
Description
PathHunter Anti-EA Antibody is an antibody against the large enzyme acceptor (EA) fragment of β-galactosidase used in an enzyme fragment complementation assay. This antibody is a mouse monoclonal of subclass IgG2a(κ). This antibody is provided as a vial of 100μg of antibody in buffer at 2mg/mL.Specifications
| 100 μg | |
| Antibodies |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.