Learn More
DiscoverX PathHunter™ Anti-EA Antibody
The PathHunter™ Anti-EA Antibody is an antibody against the large enzyme acceptor (EA) fragment of β-galactosidase. This antibody is provided as a vial of 100μg of antibody in buffer at 2mg/mL.
Brand:  DiscoverX 93-0246L
Encompass
Product Code. 17332829
  • € 1933.50 / 100µg
Description
PathHunter Anti-EA Antibody is an antibody against the large enzyme acceptor (EA) fragment of β-galactosidase used in an enzyme fragment complementation assay. This antibody is a mouse monoclonal of subclass IgG2a(κ). This antibody is provided as a vial of 100μg of antibody in buffer at 2mg/mL.
Specifications
Specifications
100 μg
Antibodies