missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ C2C12 TACR1 β-Arrestin Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ TACR1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. [ligand]) based activation of TACR1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged TACR1 and EA-tagged β-Arrestin 2. Activation of TACR1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | β-Arrestin recruitment |
| Cell Line | C2C12 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Substance P |
| Quantity | 2 Vials |
| Target Entry | TACR1 |
| Accession Number | NM_001058 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction