missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress ACVRL1/BMPR2 Dimerization Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ ACVRL1/BMPR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. GDF2/BMP9) induced dimerization of ACVRL1/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses ACVRL1 target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of ACVRL1/BMPR2 , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | ACVRL1/BMPR2 Dimerization |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | GDF-2 |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | ACVRL1/BMPR2 |
| Cell Line | U2OS |
| Target | ACVRL1/BMPR2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction