missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress EpoR-JAK2 Functional Assay
Description
The PathHunter™ EpoR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses EpoR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged EpoR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Receptor Phosphorylation |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Erythropoietin |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | EpoR-JAK2 |
| Cell Line | U2OS |
| Target | EpoR-JAK2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?