missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress IL23R/IL12RB1 Dimerization Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ IL23R/IL12RB1 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-23) induced dimerization of IL23R/IL12RB1 receptors. This product uses EFC technology. The cell line included overexpresses IL23R target protein fused to PK alongside IL12RB1 target protein fused to EA. Upon ligand-induced dimerization of IL23R/IL12RB1 , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | IL23R/IL12RB1 Dimerization |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | IL-23 |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | IL23R/IL12RB1 |
| Cell Line | U2OS |
| Target | IL23R/IL12RB1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction