missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ Sargramostim Bioassay Kit
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ Sargramostim Bioassay kit provides an easy-to-use cell based assay to determine Sargramostim potency and detect neutralizing antibodies. This bioassay assesses GM-CSF or sargramostim-induced dimerization of CSF2RB and CSF2RA receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside CSF2RA target protein fused to EA. Upon ligand-induced dimerization of CSF2RB and CSF2RA , detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | SH2 Recruitment |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | CSF2RB/CSF2R |
| Cell Line | U2OS |
| Target | CSF2RB/CSF2R |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction