missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS IL12RB1/IL12RB2 Dimerization Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ IL12RB1/IL12RB2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-12) induced dimerization of IL12RB1/IL12RB2 receptors. This product uses EFC technology. The cell line included overexpresses IL12RB2 target protein fused to PK alongside IL12RB1 target protein fused to EA. Upon ligand-induced dimerization of IL12RB1/IL12RB2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Dimerization |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | IL-12 |
| Quantity | 2 Vials |
| Target Entry | IL12RB1/IL12RB2 |
| Accession Number | NM_005535 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction