missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS IL2RB/IL2RG/IL2RA Dimerization Bioassay Kit
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ IL2RB/IL2RG/IL2RA Bioassay kit provides an easy-to-use cell based assay to determine IL-2 potency and detect neutralizing antibodies. This bioassay assesses IL2-induced multimerization of IL2RB/IL2RG/IL2RA receptors. This product uses EFC technology. The cell line included overexpresses IL2RB target protein fused to PK alongside IL2RG target protein fused to EA and untagged IL2RA. Upon ligand-induced multimerization of IL2RB/IL2RG/IL2RA , detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Dimerization |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | IL2 |
| Cell Line | U2OS |
| Target | IL2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction