missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS MCHR1 Bioassay Kit
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ U2OS MCHR1 Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay asseses ligand (e.g. Recombinant Human PDGF-BB) based on activation of MCHR1 GPCR activity via detection of SH2 Recruitment. The included cell line overexpresses ProLink-tagged MCHR1 and EA-tagged. Activation of MCHR1 stimulates the recruitment of and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | SH2 Recruitment |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 200 dp (2 x 96-well) |
| Cell Line | U2OS |
| Target | MCHR1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction